missing translation for 'onlineSavingsMsg'
Learn More

SUPT5H Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£145.00 - £417.00

Specifications

Antigen SUPT5H
Dilution Western Blot 1:200 - 1:2000, ELISA, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30229294
View Documents
Novus Biologicals
NBP3-38598-100ul
100 μL
£417.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30230391
View Documents
Novus Biologicals
NBP3-38598-20ul
20 μL
£145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SUPT5H Polyclonal antibody specifically detects SUPT5H in Human,Mouse samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications

Specifications

SUPT5H
ELISA, Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Human, Mouse
DRB sensitivity-inducing factor 160 kDa subunit, DRB sensitivity-inducing factor large subunit, DSIF large subunit, DSIF p160, FLJ34157, SPT5hSPT5, SPT5HTat-cotransactivator 1 protein, suppressor of Ty (S.cerevisiae) 5 homolog, suppressor of Ty 5 homolog (S. cerevisiae), Tat-CT1, Tat-CT1 protein, transcription elongation factor SPT5
Recombinant fusion protein containing a sequence corresponding to amino acids 125-425 of human SUPT5H (NP_003160.2).,, Sequence:, LQNLWRDQREEELGEYYMKKYAKSSVGETVYGGSDELSDDITQQQLLPGVKDPNLWTVKCKIGEERATAISLMRKFIAYQFTDTPLQIKSVVAPEHVKGYIYVEAYKQTHVKQAIEGVGNLRLGYWNQQMVPIKEMTDVLKVVKEVANLKPKSWVRLKRGIYKDDIAQVDYVEPSQNTISLKMIPRIDYDRIKARMSLKDWFAKRKKFKRPPQRLFDAEKIRSLGGDVASDGDFLIFEGNRYSRKGFLFKSFAMSAVITEGVKPTLSELEKFEDQPEGIDLEVVTESTGKEREHNFQPGDN
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot 1:200 - 1:2000, ELISA, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells
Polyclonal
Purified
RUO
PBS (pH 7.3), 50% glycerol
6829
IgG
Affinity purified
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.