missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUPT5H Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38598-100ul
This item is not returnable.
View return policy
Description
SUPT5H Polyclonal antibody specifically detects SUPT5H in Human,Mouse samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications
| SUPT5H | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, ELISA, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells | |
| DRB sensitivity-inducing factor 160 kDa subunit, DRB sensitivity-inducing factor large subunit, DSIF large subunit, DSIF p160, FLJ34157, SPT5hSPT5, SPT5HTat-cotransactivator 1 protein, suppressor of Ty (S.cerevisiae) 5 homolog, suppressor of Ty 5 homolog (S. cerevisiae), Tat-CT1, Tat-CT1 protein, transcription elongation factor SPT5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 125-425 of human SUPT5H (NP_003160.2).,, Sequence:, LQNLWRDQREEELGEYYMKKYAKSSVGETVYGGSDELSDDITQQQLLPGVKDPNLWTVKCKIGEERATAISLMRKFIAYQFTDTPLQIKSVVAPEHVKGYIYVEAYKQTHVKQAIEGVGNLRLGYWNQQMVPIKEMTDVLKVVKEVANLKPKSWVRLKRGIYKDDIAQVDYVEPSQNTISLKMIPRIDYDRIKARMSLKDWFAKRKKFKRPPQRLFDAEKIRSLGGDVASDGDFLIFEGNRYSRKGFLFKSFAMSAVITEGVKPTLSELEKFEDQPEGIDLEVVTESTGKEREHNFQPGDN | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6829 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction