missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Supor™ 450 Membrane Disc Filters

Product Code. 17134491
Change view
Click to view available options
Diameter (Metric):
13 mm
47 mm
50 mm
90 mm
102 mm
142 mm
293 mm
Surface Type:
Plain
Tabbed
Unit Size:
Pack of 100
Pack of 25
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Surface Type unitSize
17134491 47 mm Plain Pack of 100
17164491 142 mm Tabbed Pack of 25
17124491 13 mm Plain Pack of 100
17184491 293 mm Tabbed Pack of 25
17194571 293 mm Plain Pack of 25
17114501 90 mm Plain Pack of 100
17144491 50 mm Plain Pack of 100
17154491 102 mm Plain Pack of 100
17184571 142 mm Plain Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 17134491 Supplier Cytiva (formerly Pall Lab) Supplier No. 60173
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

High flow rate membrane optimized for biological, pharmaceutical, and sterilizing filtration requirements.

  • Fast filtration with superior flow rates and high throughputs
  • Low protein binding and extensive drug compatibility for critical applications
  • Saves time and money with fewer filter changes per sample volume
  • 142 and 293 mm discs feature printed tab for instant recognition of pore size and lot number
  • Designed for laboratory applications only
  • This product is not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY

Specifications

Material PES
Pore Size 0.45 μm
Sterility Non-sterile
Surface Type Plain
Thickness (Metric) 140 μm
Diameter (Metric) 47 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.