Learn More
Bollé Safety SUPERBLAST Multi-Use Autoclavable Goggle
Click to view available options
Frame Color:
Translucent
Unit Size:
1 pair
Description
Equipped with our exclusive anti-fog and anti-scratch PLATINUM coating on both sides of the lens, our range of autoclavable multi-use goggles are resistant to high temperature and still fully functional after several autoclaving cycles. Our goggles’ designs fit over any prescription glasses, ensuring improved eye protection and vision. Overflow chute for liquids Upper ventilation / sealed in the lower part Compatible with a half-mask respirator PLATINUM coating : anti-scratch & anti-fog
Specifications
Specifications
| Lens Type | Platinum |
| Product Type | Goggles |
| Frame Color | Translucent |
| Disposable | Reusable |
| Lens Material | Polycarbonate |
| Frame Material | TPR |
| Frame Type | Direct ventilated on the upper part and sealed on the bottom |
| Sterility | Sterile |