missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Flexible and adjustable nose bridge
- UV protection
- Anti-fog
- Scratch-resistant
Specifications
Specifications
| Certifications/Compliance | EN 166 (personal eye protection) and EN 170 (UV filters) |
| Lens Coating | Anti-fog, Scratch-resistant |
| Color | Light Blue, White |
| Filter Type | 2C |
| Lens Tint | Clear |
| Product Type | Safety Glasses |
| Frame Color | White, Blue |
| No. per Pack | 10/Pk. |
| Size | Regular |
| Temple Style | Soft temples |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?