missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUMO Activating Enzyme E1 (SAE1) Antibody (9D8), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | SUMO Activating Enzyme E1 (SAE1) |
|---|---|
| Clone | 9D8 |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
| Applications | Western Blot, ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30038871
|
Novus Biologicals
NBP3-26336-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30032478
|
Novus Biologicals
NBP3-26336-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SUMO Activating Enzyme E1 (SAE1) Monoclonal antibody specifically detects SUMO Activating Enzyme E1 (SAE1) in Human samples. It is validated for Western Blot, ELISA, ImmunocytochemistrySpecifications
| SUMO Activating Enzyme E1 (SAE1) | |
| Western Blot 1:500-1:5000, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AOS1activator of SUMO1, FLJ3091, HSPC140, Sua1, SUA1sentrin/SUMO-activating protein AOS1, SUMO-1 activating enzyme E1 N subunit, SUMO1 activating enzyme subunit 1, SUMO-1 activating enzyme subunit 1, SUMO-activating enzyme subunit 1, Ubiquitin-like 1-activating enzyme E1A, ubiquitin-like protein SUMO-1 activating enzyme, UBLE1A | |
| A synthesized peptide derived from Human SUMO Activating Enzyme E1 (SAE1) [UniProt Q9UBE0] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 9D8 | |
| Western Blot, ELISA, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific, Stem Cell Markers | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 10055 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title