missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUMO Activating Enzyme E1 (SAE1) Antibody (9D8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26336-100ul
This item is not returnable.
View return policy
Description
SUMO Activating Enzyme E1 (SAE1) Monoclonal antibody specifically detects SUMO Activating Enzyme E1 (SAE1) in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry
Specifications
| SUMO Activating Enzyme E1 (SAE1) | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry | |
| 9D8 | |
| Western Blot 1:500-1:5000, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| AOS1activator of SUMO1, FLJ3091, HSPC140, Sua1, SUA1sentrin/SUMO-activating protein AOS1, SUMO-1 activating enzyme E1 N subunit, SUMO1 activating enzyme subunit 1, SUMO-1 activating enzyme subunit 1, SUMO-activating enzyme subunit 1, Ubiquitin-like 1-activating enzyme E1A, ubiquitin-like protein SUMO-1 activating enzyme, UBLE1A | |
| A synthesized peptide derived from Human SUMO Activating Enzyme E1 (SAE1) [UniProt Q9UBE0] | |
| 100 μL | |
| Phospho Specific, Stem Cell Markers | |
| 10055 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction