missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sulindac

Product Code. 15404299
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15404299 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15404299 Supplier Thermo Scientific Chemicals Supplier No. J61772.06
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 38194-50-2 | C20H17FO3S, C20H17FO3S | 356.41 g/mol

Sulindac inhibits the NF-κB pathway. Sulindac is a non-steroidal anti-inflammatory drug. Sulindac reduces the level and activity of several cyclin-dependent kinases and induces apoptosis. It is an antitumor agent. Sulindac is an inhibitor of Cox-1 and Cox-2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase-1 and induces apoptosis in HT-29 cells.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38194-50-2
Molecular Formula C20H17FO3S
Synonym cis-5-Fluoro-2-methyl-1-[4-(methylsulfinyl)benzylidene]indene-3-acetic acid; MK-231

Specifications

Quantity 5 g
Description Yellow
Content And Storage -30°C to -10°C
Format Powder
For Use With (Application) A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase-1 and induces apoptosis in HT-29 cells
Formula Weight 356.41g/mol
Melting Point 182°C to 185°C
Product Type Signal Transduction Reagent
Form Powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.