missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfotyrosine Antibody (7C5), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£156.00 - £524.00
Specifications
| Antigen | Sulfotyrosine |
|---|---|
| Clone | 7C5 |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18245833
|
Novus Biologicals
NBP2-59384 |
100 μg |
£524.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18604488
|
Novus Biologicals
NBP2-59384-25ug |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sulfotyrosine Monoclonal specifically detects Sulfotyrosine in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| Sulfotyrosine | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG1 | |
| Protein G purified |
| 7C5 | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Synthetic Sulfotyrosine conjugated to Keyhole Limpet Hemocyanin (KLH). | |
| Primary | |
| Specific for sulfated tyrosine residues on various proteins. Does not detect free Sulfotyrosine. Does not cross-react with phosphotyrosine. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title