missing translation for 'onlineSavingsMsg'
Learn More

Sulfotyrosine Antibody (7C5), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication

Brand:  Novus Biologicals NBP2-59384-25ug

 View more versions of this product

Product Code. 18604488

  • £156.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sulfotyrosine Monoclonal specifically detects Sulfotyrosine in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sulfotyrosine
Monoclonal
Unconjugated
PBS (pH 7.4), 50% glycerol with 0.09% Sodium Azide
Synthetic Sulfotyrosine conjugated to Keyhole Limpet Hemocyanin (KLH).
25 μg
Primary
Store at -20°C.
IgG1
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
7C5
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50
Mouse
Protein G purified
RUO
Specific for sulfated tyrosine residues on various proteins. Does not detect free Sulfotyrosine. Does not cross-react with phosphotyrosine.
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only