missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Sulforhodamine 101
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Sulforhodamine 101 is used extensively for investigating neuronal morphology and acts as a cell type-selective fluorescent marker of astrocytes both in vivo and in slice preparations.
Specifications
Specifications
| Content And Storage | -20°C, protect from light ∣ The compound is unstable in solutions, freshly prepared is recommended. |
| Product Type | Dye Reagents |
| Quantity | 25 mg |
| Shipping Condition | Room temperature |
| Solubility | DMSO : 50 mg/mL (82.41 mM; Need ultrasonic) ∣ H2O : 8.33 mg/mL (13.73 mM; Need ultrasonic) |
| Molecular Weight (g/mol) | 606.71 g/mol |
| CAS | 60311-02-6 |
| Synonym | SR101 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction