missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SulfoLink™ Coupling Resin

Product Code. 11896774 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Unit Size:
10mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11896774 50 mL 50mL
10485505 10 mL 10mL
11796406 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11896774 Supplier Thermo Scientific™ Supplier No. 20402
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Covalently immobilize cysteine-peptides or proteins (sulfhydryl-ligands) for affinity purification using iodoacetyl-activated agarose beads and columns.

Thermo Scientific™ SulfoLink Coupling Resin is iodoacetyl-activated for the covalent immobilization of sulfhydryl-containing peptides or proteins for affinity purification.

SulfoLink Coupling Resin is porous, crosslinked 6% beaded agarose that has been activated with iodoacetyl groups. When incubated with a solution of peptide or protein that contains reduced cysteine residues, the iodoacetyl groups react specifically and efficiently with the exposed sulfhydryls (-SH) to form covalent and irreversible thioether bonds that permanently attach the peptide or protein to the resin. The result is a custom-made affinity resin for purification of antibodies, antigens and other molecules of interest.

Complete SulfoLink Immobilization Kits are available for peptides and proteins. Each kit contains all the reagents for preparing the respective peptide or protein sample and five columns containing 2mL of SulfoLink Resin that can be used in either gravity-flow or centrifuge format for efficient coupling reactions and multiple cycles of affinity purification. A single-column Trial Kit for use with either peptides or proteins is also available, or the SulfoLink Coupling Resin can be purchased separately.

Highlights:

  • Specific conjugation through sulfhydryl (-SH) groups – the iodoacetyl groups react specifically with sulhydryls to form irreversible thioether bonds

  • Separate kits optimized for peptides or proteins – kits include optimized reagents for preparing peptide or protein samples for efficient immobilization

  • Fast – spin columns increase protocol speed; prepare and couple samples in 2 hours (peptides) to 3.5 hours (proteins)

  • Flexible coupling conditions – use pH 7.5 to 9.0 aqueous buffers, organic solvent (e.g., 20% DMSO) or denaturant (guanidine·HCl), as needed for protein or peptide solubility during coupling reaction

  • Easy-to-follow instructions – streamlined protocols for sample preparation, immobilization and affinity purification

  • High capacity – immobilize 1 to 2mg peptide or 2 to 20mg protein per 2mL column of SulfoLink Coupling Resin

Includes:

Kits contain SulfoLink Resin Columns (2mL), protein or peptide preparation reagents (e.g., reducing agent), coupling buffer, wash solution, L-cysteine solution (for quenching) and desalting columns (protein kit only)

Recommended for:

Immobilize peptides having terminal cysteine residues to purify antibodies that were generated against peptide immunogens prepared by maleimide conjugation; Immobilize antibodies in oriented manner through hinge-region sulfhydryls to ensure that antigen binding sites are not sterically hindered for antigen affinity purification

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Upon receipt store product in the dark at 4°C. Product is shipped at ambient temperature.
Form Liquid Suspension
Product Line SulfoLink
Quantity 50 mL
Type Resin
Stationary Phase Haloacetyl
Sufficient For 1 mg Peptide or 10 mg Antibody per mL of Resin
Format Bottle
Column Type Affinity, Agarose Resin

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.