missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sulfo-SBED Biotin Label Transfer Reagent, No-Weigh™ Format

Product Code. 16317591 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mg
8 x 1 mg
10 x 1 mg
Unit Size:
10mg
Pack of 10
Pack of 8
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16317591 10 x 1 mg Pack of 10
10127883 8 x 1 mg Pack of 8
11899240 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16317591 Supplier Thermo Scientific™ Supplier No. A39260

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemically tag purified proteins at primary amines to covalently crosslink and transfer biotinylation to protein interaction binding partners.

Thermo Scientific™ Pierce Sulfo-SBED Biotin Label Transfer Reagent is a multifunctional reagent for labeling a purified protein and then covalently transferring the attached biotin tag onto specific interactors of that protein.

  • Multifunctional reagent for labeling a purified protein and then covalently transferring the attached biotin tag onto specific interactors of that protein.
  • Heterobifunctional chemical crosslinker capable of covalently attaching to primary amines at one end and to nearly any protein functional group at the other end.
  • Includes a biotin group and a cleavable disulfide spacer arm.
  • Together these features allow one to sequentially crosslink interacting proteins and transfer the biotin affinity tag from one protein (i.e., a purified bait protein) to another (possibly unknown prey protein).
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 x 1 mg
Product Type Sulfo-SBED Biotin Label Transfer Reagent
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Methanol, Water
Content And Storage Store at 4°C.
Chemical Reactivity Amine-to-Amine
Labeling Method Chemical Labeling, Label Transfer
Shipping Condition Qualified for Ambient or Wet Ice
Reactive Moiety NHS Ester, Alkyne
Label or Dye Biotin
Label Type Biotin and Other Haptens
Product Line Pierce
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.