missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Subcellular Protein Fractionation Kit

Product Code. 18715143 Shop All Bio Techne Products
Click to view available options
Quantity:
50 Assays
Unit Size:
50 assays
This item is not returnable. View return policy

Product Code. 18715143

Brand: Novus Biologicals™ NBP247659

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed to provide reproducible extraction of four subcellular protein fractions (cytosol, nucleus, membrane/particulate, and cytoskeletal fractions) from a single mammalian sample; the percentage of proteins in the different fractions are cell-dependent, usually in the range of approx.20-30% cytosol, approx.25-35% membrane (organelles), approx.15-25% Nuclei, approx.15-25% insoluble

In Western analysis, if a membrane protein appeares mainly in the nuclear fraction, the membrane extraction step may not be sufficient. First, make sure to add Membrane Extraction Buffer B into the extraction, which contain NP-40 detergent. If it still shows the same result, increase incubation time from 1 min to 5 min after adding MEB-B. NP-40 should extract membrane from most of cells.
TRUSTED_SUSTAINABILITY

Specifications

Includes Protein Fractionation Kit
Product Type Subcellular Fractions
Target Proteins Subcellular Protein Fractionation
Content And Storage Store at -20°C Avoid freeze-thaw cycles
For Use With (Application) 1-D or 2-D gel, enzyme activity assays, gel supershift assay, and Western blotting; Subcellular Protein Fractionation: The Cell Fractionation Kit can be used in 1-D or 2-D gel, enzyme activity assays, gel shift assay, and Western blotting.
Quantity 50 Assays
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.