missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ SUB-GEL Teaching Horizontal Gel Tank Package

Product Code. p-6827066 Shop All Fisherbrand Products
Click to view available options
Type:
6X Mini Classroom Workstation
Teaching Gel Tank, 4 UV trays
Teaching Gel Tank, 6 UV trays
Includes:
Four 70mm x 70mm UV trays, eight 8 sample combs, eight casting dams, loading guides and cables
Four 70mm x 70mm UV trays, eight 8 sample combs, eight casting dams, loading guides and cables; Plus power supply (Cat. No 12643546)
Four SUB-GEL 6X Mini units (Cat. No 15321357) and power supply (Cat. No 12613546)
Six 70mm x 70mm UV trays, twelve 8 sample combs, twelve casting dams, loading guides and cables
Six 70mm x 70mm UV trays, twelve 8 sample combs, twelve casting dams, loading guides and cables; Plus power supply (Cat. No 12643546)
Unit Size:
Each

Product Code. 15341357

Only null left
Add to basket
Add to basket

SUB-GEL Teaching gel tanks are ideal for demonstration of horizontal gel electrophoresis, helps teachers to demonstrate horizontal gel electrophoresis to many students using just single gel tank and power supply.

  • High quality injection moulded construction
  • Leak free ‘Plug-and-Go’ casting dams
  • UV transparent gel trays
  • Height adjustable combs
  • Low gel and buffer volumes
  • Cassette-style electrodes - difficult to break and easy to change
  • Each tank is available with or without power supply

Specifications

Description Standard combs: Eight x 8 sample, 1mm MC (2 combs per gel)
Device Holds Maximum Gel throughput: Four 70mm x 70mm mini Gels
For Use With (Application) Demonstrating Horizontal Gel Electrophoresis to Students
Type Teaching Gel Tank, 4 UV trays
Gel Dimensions 7 x 7 cm (2.7 x 2.7 in.)
Includes Four 70mm x 70mm UV trays, eight 8 sample combs, eight casting dams, loading guides and cables
Sample Size 64 sample with standard comb option, 128 samples using optional 16 sample combs
Volume (Metric) Buffer 500 mL
Dimensions (L x W x H) 26.5 x 17.5 x 9 cm (10.4 x 6.8 x 3.5 in.)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.