missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Fisherbrand™ SUB-GEL Teaching Horizontal Gel Tank Package
Shop alle Fisherbrand produkter
missing translation for 'orderingAttributeHoverText'
Type:
6X Mini Classroom Workstation
Teaching Gel Tank, 4 UV trays
Teaching Gel Tank, 6 UV trays
Includes:
Four 70mm x 70mm UV trays, eight 8 sample combs, eight casting dams, loading guides and cables
Four 70mm x 70mm UV trays, eight 8 sample combs, eight casting dams, loading guides and cables; Plus power supply (Cat. No 12643546)
Four SUB-GEL 6X Mini units (Cat. No 15321357) and power supply (Cat. No 12613546)
Six 70mm x 70mm UV trays, twelve 8 sample combs, twelve casting dams, loading guides and cables
Six 70mm x 70mm UV trays, twelve 8 sample combs, twelve casting dams, loading guides and cables; Plus power supply (Cat. No 12643546)
Förpackningsstorlek
Each
Tekniske data
Tekniske data
| Description | Standard combs: Eight x 8 sample, 1mm MC (2 combs per gel) |
| Device Holds | Maximum Gel throughput: Four 70mm x 70mm mini Gels |
| For Use With (Application) | Demonstrating Horizontal Gel Electrophoresis to Students |
| Type | Teaching Gel Tank, 4 UV trays |
| Gel Dimensions | 7 x 7 cm (2.7 x 2.7 in.) |
| Includes | Four 70mm x 70mm UV trays, eight 8 sample combs, eight casting dams, loading guides and cables |
| Sample Size | 64 sample with standard comb option, 128 samples using optional 16 sample combs |
| Volume (Metric) Buffer | 500 mL |
| Dimensions (L x W x H) | 26.5 x 17.5 x 9 cm (10.4 x 6.8 x 3.5 in.) |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion