missing translation for 'onlineSavingsMsg'
Learn More

STX1A Rabbit, anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen STX1A
Concentration 350 μg/mL
Applications Western Blot, ELISA
Form Liquid
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18060306
View Documents
Proteintech
55164-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18057795
View Documents
Proteintech
55164-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The STX1A antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human STX1A. This antibody recognizes human antigen. The STX1A antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Specifications

STX1A
Western Blot, ELISA
Rabbit
6804
IgG
Store at -20°C. Stable for one year after shipment.
350 μg/mL
Liquid
Human
HPC 1, Neuron specific antigen HPC 1, p35 1, STX1, STX1A, Syntaxin 1A, syntaxin 1A (brain)
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.