missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STX1A Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55164-1-AP-20UL
This item is not returnable.
View return policy
Description
The STX1A antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human STX1A. This antibody recognizes human antigen. The STX1A antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| STX1A | |
| 350 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| HPC 1, Neuron specific antigen HPC 1, p35 1, STX1, STX1A, Syntaxin 1A, syntaxin 1A (brain) | |
| 20 μL | |
| 6804 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction