missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STX16 Recombinant Rabbit Monoclonal Antibody (23GB4655), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | STX16 |
|---|---|
| Clone | 23GB4655 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087992
![]() |
Invitrogen™
MA553782 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089582
|
Invitrogen™
MA553783 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
STX16 Recombinant Monoclonal antibody specifically detects STX16 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| STX16 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| O14662 | |
| 8675 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4655 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 4930401D03; 5430410K23Rik; 6330500A18Rik; AI648908; AW553605; CG1467; CG1467-PA; Dm Syx16; Dmel\CG1467; Dmel_CG1467; DmSyx16; dSyntaxin16; dSyx 16; dSyx16; MGC90328; RP11-261P9.3; STX16; Syn16; syntaxin; syntaxin 16; Syntaxin16; syntaxin-16; Syx; SYX16; Syx16-PA | |
| A synthesized peptide derived from human Syntaxin 16 (50-90AA). | |
| Primary | |
| -20°C | |
| STX16 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title