missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STX16 Recombinant Rabbit Monoclonal Antibody (23GB4655), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553783
This item is not returnable.
View return policy
Description
This gene encodes a protein that is a member of the syntaxin or t-SNARE (target-SNAP receptor) family. These proteins are found on cell membranes and serve as the targets for V-SNARES (vesicle-SNAP receptors) permitting specific synaptic vesicle docking and fusion. A microdeletion in the region of chromosome 20 where this gene is located has been associated with pseudohypoparathyroidism type Ib. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| STX16 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O14662 | |
| STX16 | |
| A synthesized peptide derived from human Syntaxin 16 (50-90AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4655 | |
| Unconjugated | |
| STX16 | |
| 4930401D03; 5430410K23Rik; 6330500A18Rik; AI648908; AW553605; CG1467; CG1467-PA; Dm Syx16; Dmel\CG1467; Dmel_CG1467; DmSyx16; dSyntaxin16; dSyx 16; dSyx16; MGC90328; RP11-261P9.3; STX16; Syn16; syntaxin; syntaxin 16; Syntaxin16; syntaxin-16; Syx; SYX16; Syx16-PA | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 8675 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction