missing translation for 'onlineSavingsMsg'
Learn More

Strontium neodecanoate, Sr 16-21%, Thermo Scientific™

Product Code. 11344477
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344477 10 g 10g
11364477 2 g 2g
11354477 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11344477 Supplier Thermo Scientific Alfa Aesar Supplier No. 039574.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106705-37-7
Molecular Formula C20H38O4Sr
Molecular Weight (g/mol) 430.14
MDL Number MFCD00070439
InChI Key CSQXYUPPOWGAIS-UHFFFAOYSA-L
Synonym strontium neodecanoate, sr, strontium bis 7,7-dimethyloctanoate, strontium 2+ bis 7,7-dimethyloctanoate
PubChem CID 19359001
IUPAC Name strontium;7,7-dimethyloctanoate
SMILES CC(C)(C)CCCCCC(=O)[O-].CC(C)(C)CCCCCC(=O)[O-].[Sr+2]

Specifications

Quantity 10 g
Formula Weight 430.1
Concentration or Composition (by Analyte or Components) Sr 16 to 21%
Physical Form Powder
Chemical Name or Material Strontium neodecanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.