accessibility menu, dialog, popup

UserName

CAS RN 106705-37-7

H3CCH3H3COOH3CCH3H3COOSr

CAS RN 106705-37-7

IUPAC Name: strontium(2+) bis(7,7-dimethyloctanoate)
Synonyms: strontium(2+) bis(7,7-dimethyloctanoate)
Molecular Weight (g/mol): 430.14
Molecular Formula: C20H38O4Sr
InChi Key: CSQXYUPPOWGAIS-UHFFFAOYSA-L
SMILES: [Sr++].CC(C)(C)CCCCCC([O-])=O.CC(C)(C)CCCCCC([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results