missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Striatin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
£183.00 - £426.00
Specifications
| Antigen | Striatin |
|---|---|
| Dilution | Western Blot 1:500-1:5000, Immunocytochemistry/Immunofluorescence 1:5000, Immunoprecipitation 1:10-1:500 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18251370
|
Novus Biologicals
NB110-74571SS |
0.025 mL |
£183.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18725393
|
Novus Biologicals
NB110-74571 |
0.1 mL |
£426.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Striatin Polyclonal specifically detects Striatin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| Striatin | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication, Neuroscience, Neurotransmission, Signal Transduction | |
| Whole antisera with 0.02% Sodium Azide | |
| MGC125642, SG2NA, striatin, striatin, calmodulin binding protein, striatin, calmodulin-binding protein | |
| STRN | |
| IgG | |
| Unpurified | |
| This antibody specific for Striatin. It does not cross-react with SG2NA. |
| Western Blot 1:500-1:5000, Immunocytochemistry/Immunofluorescence 1:5000, Immunoprecipitation 1:10-1:500 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat | |
| Q13033 | |
| 6801 | |
| Synthetic peptide made to an internal portion of the human Striatin protein (within residues 350-400). [Swiss-Prot# O43815] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 110 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title