missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Striatin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NB110-74571SS
This item is not returnable.
View return policy
Description
Striatin Polyclonal specifically detects Striatin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| Striatin | |
| Polyclonal | |
| Western Blot 1:500-1:5000, Immunocytochemistry/Immunofluorescence 1:5000, Immunoprecipitation 1:10-1:500 | |
| Q13033 | |
| STRN | |
| Synthetic peptide made to an internal portion of the human Striatin protein (within residues 350-400). [Swiss-Prot# O43815] | |
| Unpurified | |
| RUO | |
| Primary | |
| This antibody specific for Striatin. It does not cross-react with SG2NA. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Whole antisera with 0.02% Sodium Azide | |
| MGC125642, SG2NA, striatin, striatin, calmodulin binding protein, striatin, calmodulin-binding protein | |
| Rabbit | |
| 110 kDa | |
| 0.025 mL | |
| Cell Biology, Cell Cycle and Replication, Neuroscience, Neurotransmission, Signal Transduction | |
| 6801 | |
| Human, Mouse, Rat | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction