missing translation for 'onlineSavingsMsg'
Learn More

Cytiva StrepTrap™ HP Prepacked Columns

Product Code. 10298754 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 1mL
1 x 5mL
5 x 1mL
5 x 5mL
Flow Rate:
<20 mL/min.
<4 mL/min.
Content And Storage:
2° to 8°C
4° to 8°C
Unit Size:
1mL
Each
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Flow Rate Content And Storage unitSize
10298754 1 x 5mL <20 mL/min. 4° to 8°C Each
11540654 5 x 5mL <20 mL/min. 4° to 8°C Pack of 5
10288754 5 x 1mL <4 mL/min. 2° to 8°C Pack of 5
15117956 1 x 1mL <4 mL/min. 2° to 8°C 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10298754 Supplier Cytiva Supplier No. 28907547

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purify Strep(II)-tagged recombinant proteins by affinity chromatography. Cytiva Life Sciences™ StrepTrap™ HP Prepacked Columns are packed with a robust and stable Sepharose™ based medium which results in high resolution, sharp concentrated peaks, and reproducible purifications. The binding capacity is also high.

  • Prepacked with StrepTactin Sepharose High Performance
  • Binding capacity Approx. 6 mg Strep II-tagged protein/ml medium
  • Maximum pressure: 3 bar
  • Column material: polypropylene
  • Compatible with a wide range of reducing agents, detergents, denaturants, and other additives
  • Generated with 0.5M NaOH
  • 1 and 5mL columns

Media
  • Ligand: Streptactin
  • Matrix: 6% cross-linked agarose
  • Average particle size: 34μm
  • pH stability working range: >7

1mL columns

  • Bed dimensions: 7 x 25mm
  • Flow velocity: <4mL/min.

5mL columns
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.

TRUSTED_SUSTAINABILITY

Specifications

Product Type StrepTactin Sepharose HP prepacked column
Bed Volume 1mL
Diameter (English) Inner 0.62 in.
Diameter (Metric) Inner 16mm
Quantity 1 x 5mL
Height (English) 0.98 in.
Height (Metric) 25mm
Flow Rate <20 mL/min.
Material Polypropylene
Ligand Type StrepTactin (∼5mg/mL medium)
Max. Pressure 3 bar
pH Range 7
Particle Size 34μm
Support Type 6% Highly Cross-linked Agarose
Content And Storage 4° to 8°C
Column Type Ion Exchange Column
For Use With (Application) Purification of Strep (II) -tagged recombinant Proteins by affinity chromato graphy
Show More Show Less
compliance-icons
Product Identifier
  • StrepTrap HP, 5 ml, 1 x 5 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.