missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptococcus agalactiae Monoclonal Antibody (Strep13-509.3), Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

£367.00 - £916.00

Specifications

Antigen Streptococcus agalactiae
Clone Strep13-509.3
Concentration 0.9 mg/mL
Applications ELISA
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17870446
View Documents
Invitrogen™
MA5-46098
100 μg
£367.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11553463
GREENER_CHOICE
View Documents
Invitrogen™
MA110875
500 μg
£475.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17840426
View Documents
Invitrogen™
MA5-46183
1 mg
£916.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MA1-10875 detects Streptococcus agalactiae from Strep agalactiae samples. MA1-10875 has been successfully used in ELISA applications. The MA1-10875 immunogen is UV-inactivated Strep agalactiae cells isolated from mastitic milk.

Once thought to infect only cows, where it produces mastitis, Streptococcus agalactiae is now known to be able to cause serious disease, bacteremia and meningitis, in immunocompromised individuals and in neonates. Infections in the newborn are the most common and are extremely serious. They are of two types. Early onset of infection, usually occuring within 5 days of birth, is manifested by bacteremia and pneumonia. This infection is contracted vertically, as the baby passes through the birth canal. The other type of infection is a meningitis that occurs between the 10th and 60th days of life. The origin of this infection is not known, but it is not believed to be the mother.
Specifications

Specifications

Streptococcus agalactiae
0.9 mg/mL
Monoclonal
Liquid
RUO
PBS with no preservative; pH 7.4
UV-inactivated Strep agalactiae cells isolated from mastitic milk.
Primary
-20°C, Avoid Freeze/Thaw Cycles
Strep13-509.3
ELISA
Unconjugated
Mouse
Bacteria
s agalactiae; S. agalactiae
IgG1
Protein G
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.