missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Streptococcus agalactiae Monoclonal Antibody (Strep13-509.3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA5-46183
This item is not returnable.
View return policy
Description
MA1-10875 detects Streptococcus agalactiae from Strep agalactiae samples. MA1-10875 has been successfully used in ELISA applications. The MA1-10875 immunogen is UV-inactivated Strep agalactiae cells isolated from mastitic milk.
Once thought to infect only cows, where it produces mastitis, Streptococcus agalactiae is now known to be able to cause serious disease, bacteremia and meningitis, in immunocompromised individuals and in neonates. Infections in the newborn are the most common and are extremely serious. They are of two types. Early onset of infection, usually occuring within 5 days of birth, is manifested by bacteremia and pneumonia. This infection is contracted vertically, as the baby passes through the birth canal. The other type of infection is a meningitis that occurs between the 10th and 60th days of life. The origin of this infection is not known, but it is not believed to be the mother.
Specifications
| Streptococcus agalactiae | |
| Monoclonal | |
| 0.9 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG1 |
| ELISA | |
| Strep13-509.3 | |
| Unconjugated | |
| s agalactiae; S. agalactiae | |
| UV-inactivated Strep agalactiae cells isolated from mastitic milk. | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction