missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Streptavidin eFluor™ 570

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry, immunohistochemistry, and immunocytochemistry applications. Streptavidin binds to biotin with high affinity.

Brand:  Invitrogen™ 41-4317-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15528936

  • £300.00 / 100µg
Estimated Shipment: 10-06-2024
to see stock.
Add to basket

Alert:

Contains Sodium azide


Description

Description

Specifications

Specifications

0.2 mg/mL
100 μg
SA, Sav
Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections, Immunohistochemical Staining of Frozen Tissue Sections, Immunocytochemistry
eFluor 570
Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
2°C to 8°C. Do not freeze. Light-sensitive material.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only