missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Streptavidin eFluor™ 570
Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry, immunohistochemistry, and immunocytochemistry applications. Streptavidin binds to biotin with high affinity.
£135.00 - £300.00
Specifications
Concentration | 0.2 mg/mL |
---|---|
Conjugate | eFluor 570 |
Storage Buffer | Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer |
Synonym | SA, Sav |
Content And Storage | 2°C to 8°C. Do not freeze. Light-sensitive material. |
Alert:
Contains Sodium azide
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15518936
|
Invitrogen™
41-4317-80 |
25 μg |
£135.00
25µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15528936
|
Invitrogen™
41-4317-82 |
100 μg |
£300.00
100µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
Specifications
0.2 mg/mL | |
Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer | |
2°C to 8°C. Do not freeze. Light-sensitive material. |
eFluor 570 | |
SA, Sav | |
Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections, Immunohistochemical Staining of Frozen Tissue Sections, Immunocytochemistry |
For Research Use Only