missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences Strep-Tactin™XT Superflow™ High Capacity Column
Strep-Tactin™XT Superflow™ high capacity column is a gravity flow column to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
£197.49 - £509.76
Specifications
Concentration | 6% agarose, crosslinked |
---|---|
Formulation | Ready-to-use column with Strep-Tactin™XT high capacity resin |
Content And Storage | 2°C to 8°C |
For Use With (Application) | Purification of Strep-Tag™II and Twin-Strep-Tag™ Fusion Proteins |
Type | Pre-packed Column |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
16401485
|
IBA Lifesciences
2-4034-001 |
10 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16491475
|
IBA Lifesciences
2-4031-005 |
5 x 0.2 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Strep-Tactin™XT Superflow™ high capacity columns contain a 6% agarose coupled with the streptavidin variant Strep-Tactin™XT and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins via gravity flow.
- They enable the addition of denaturing agents such as urea during the purification procedure.
- In comparison to Strep-Tactin™XT Superflow™, Strep-Tactin™XT Superflow™ high capacity has a higher density of immobilized Strep-Tactin™XT, which leads to a 3- to 5-fold higher yield of purified protein.
- As with any other Strep-Tactin™XT resins, the unspecific protein binding is extremely low, which results in an outstanding purity of the target protein.
- To reduce the waiting period during the sample and buffer application, the use of the application aid WET FRED is recommended.
Specifications
6% agarose, crosslinked | |
2°C to 8°C | |
Pre-packed Column |
Ready-to-use column with Strep-Tactin™XT high capacity resin | |
Purification of Strep-Tag™II and Twin-Strep-Tag™ Fusion Proteins |
For Research Use Only.