missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Strep-Tactin™XT Superflow™ High Capacity Column

Product Code. 16491475 Shop All IBA Lifesciences Products
Click to view available options
Capacity (Metric):
10 mL
5 x 0.2 mL
Unit Size:
Each
Pack of 5
This item is not returnable. View return policy

Product Code. 16491475

Brand: IBA Lifesciences 24031005

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Strep-Tactin™XT Superflow™ high capacity column is a gravity flow column to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins

  • Strep-Tactin™XT Superflow™ high capacity columns contain a 6% agarose coupled with the streptavidin variant Strep-Tactin™XT and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins via gravity flow.
  • They enable the addition of denaturing agents such as urea during the purification procedure.
  • In comparison to Strep-Tactin™XT Superflow™, Strep-Tactin™XT Superflow™ high capacity has a higher density of immobilized Strep-Tactin™XT, which leads to a 3- to 5-fold higher yield of purified protein.
  • As with any other Strep-Tactin™XT resins, the unspecific protein binding is extremely low, which results in an outstanding purity of the target protein.
  • To reduce the waiting period during the sample and buffer application, the use of the application aid WET FRED is recommended.

Specifications

Capacity (Metric) 5 x 0.2 mL
Concentration 6% agarose, crosslinked
Formulation Ready-to-use column with Strep-Tactin™XT high capacity resin
Kit Contents Pre-packed in buffer, pH 8.0: 100 mm Tris-HCl pH 8.0, 1 mm EDTA, 150 mm NaCl, 0.02% sodium azide, 1.5 M a mmonium sulphate
Content And Storage 2°C to 8°C
Quantity 5 x 0.2 mL
For Use With (Application) Purification of Strep-Tag™II and Twin-Strep-Tag™ Fusion Proteins
Type Pre-packed Column

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.