Learn More
IBA Lifesciences Strep-Tactin™XT Superflow™ Cartridge
Strep-Tactin™XT Superflow™ cartridge is a HPLC/FPLC cartridge to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
Brand:  IBA Lifesciences 2-4022-001
 View more versions of this product
Product Code. 15728207
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Strep-Tactin™XT Superflow™ cartridges contain a 6% agarose coupled with the streptavidin variant Strep-Tactin™XT and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins with HPLC/FPLC devices like Äkta™ systems (Cytiva).
  • In comparison to Strep-Tactin™ Superflow™, Strep-Tactin™XT Superflow™ has a higher affinity for Strep-tag™ fusion proteins, which leads to an improved immobilization and purification.
  • In addition, Strep-Tactin™XT Superflow™ allows the application of denaturing agents such as urea during the purification procedure.
  • As with any other Strep-Tactin™XT resins, the unspecific protein binding is extremely low, which results in an outstanding purity of the target protein. 
Specifications
Specifications
5.7 mg protein/ mL resin
1 Cartridge
Chromatpgraphy workstations with 10 to 32 connections
1 x 5 mL
Protein Purification