Learn More
IBA GmbH™ Strep-Tactin™XT Starter Kit
Product Code. 15697028 Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. 15697028
Brand: IBA GmbH™ 24998000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
IBA GmbH™ Strep-Tactin™XT Starter Kit is a comprehensive kit for Strep-tag protein purification and detection.
  • The Strep-Tactin™XT Starter Kit contains all essential reagents required for purification and detection of Strep-tagII™ and Twin-Strep-tag™ proteins.
  • The kit contains all reagents sufficient for 8 applications
  • 1x 1 mL Strep-Tactin™XT Superflow™ gravity flow column
  • Strep-Tactin™/ Strep-Tactin™XT Wash Buffer (1x Buffer W)
  • Strep-Tactin™XT Elution Buffer with Biotin for displacing the Twin-Strep-tag™ protein from the column (1x Buffer BXT)
  • Strep-Tactin™ horse radish peroxidase (HRP) conjugate for Western blot detection
Specifications
Kit Contents Gravity flow column, wash buffer, elution buffer
Quantity 1 Kit