missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Strep-Tactin™XT Conjugate
GREENER_CHOICE

Product Code. 16401475 Shop All IBA Lifesciences Products
Click to view available options
Label or Dye:
DY-488
DY-549
DY-649
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 16401475

Brand: IBA Lifesciences 21562050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Strep-Tactin™XT conjugates with fluorescent label to detect Strep-tag™II and Twin-Strep-tag™ fusion proteins

Strep-Tactin™XT is a streptavidin variant, which binds to the Strep-tag™II and Twin-Strep-tag™ with high affinity and specificity.

It is available as conjugate with fluorescent dyes (DY-488, DY-549, DY-649) for fast and directed detection of N-terminally, C-terminally as well as internally tagged Strep-tag™II or Twin-Strep-tag™ fusion proteins in applications like ELISA, FACS, immunohistochemistry, and immunocytochemistry.

The absorption/emission maxima of the fluorescent dyes are:
• DY-488: 493/518 nm
• DY-549: 550-560/575 nm
• DY-649: 655/676 nm

Specifications

Capacity (Metric) 0.10 mL
Concentration 0.5 mg/mL
Formulation Strep-Tactin™XT conjugated with DY-488
Kit Contents Solution in PBS; 0.01 % NaN3; 1 mg/ mL BSA
Content And Storage Undiluted: 2°C to 8°C, protect from light; Diluted: -25°C to -15°C
Quantity 50 μg
For Use With (Application) ELISA, FACS, western blot, immunofluorescence
Target Strep-tag™II and Twin-Strep-tag™
Type Conjugate
Label or Dye DY-488

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.