missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences Strep-Tactin™XT 4Flow™ Starter Kit
Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Description
- Strep-Tactin™XT 4Flow™ Starter Kit contains all essential reagents required for purification and detection of Strep-tag™II and Twin-Strep-tag™ fusion proteins.
- It provides a Strep-Tactin™XT 4Flow™ column, Buffer W, Buffer BXT, and Buffer XT-R for purification as well as Strep-Tactin™ HRP for subsequent western blot detection.
- Strep-Tactin™XT 4Flow™ columns contain a 4% agarose coupled with the streptavidin variant Strep-Tactin™XT.
- Due to the broad pH compatibility, excellent pressure stability, outstanding target purity (> 95%), and highest binding affinity, the resin is recommended for all protein classes, especially for low abundant or challenging proteins.
- The low concentrated agarose enables the simple attachment of larger proteins to the stationary phase, making it particularly suitable for purification of high-molecular-weight targets.
- In addition, Strep-Tactin™XT 4Flow™ allows the application of denaturing agents such as urea.
- To reduce the waiting period during the sample and buffer application, the use of the application aid WET FRED is recommended.
Specifications
Specifications
| Matrix | 4% agarose, crosslinked |
| Quantity | 1 Kit |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction