missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Strep-Tactin™ TACS Agarose

Product Code. 17247750 Shop All IBA Lifesciences Products
Click to view available options
Capacity (Metric):
4 mL
20 mL
50 mL
Unit Size:
20mL
4mL
50mL
This item is not returnable. View return policy

Product Code. 17247750

Brand: IBA Lifesciences 66350002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

50% suspension for traceless affinity cell and exosome isolation

  • Strep-Tactin™ TACS Agarose contains agarose macrobeads coated with the streptavidin variant Strep-Tactin™.
  • A special crosslinking permits the use for cell isolation and depletion approaches.
  • It was developed for column-based Traceless Affinity Cell Selection (Fab-TACS™/Nano-TACS™).
  • In this approach, Fab fragments fused to a Twin-Strep-tag™ (Fab-Streps) or nanobodies (Nano-Streps) are bound to Strep-Tactin™ on the agarose matrix.
  • After specifically capturing cells, Fab-/Nano-Streps are released from the Strep-Tactin™ Agarose by biotin addition.
  • In this way, the eluted cells are ready for further downstream applications.
  • The same principle is applicable for exosome isolation (Fab-TACS™ for exosomes)
  • Alternatively to Fab-Streps/Nano-Streps, it is possible to bind Twin-Strep-tagged, desthiobiotinylated or biotinylated antibodies (irreversible binding) to this resin.
  • This makes Strep-Tactin™ TACS Agarose also suitable for negative cell selection or depletion approaches.
  • The suspension can be used for packing of gravity flow columns since the resin is optimized for column affinity chromatography.
  • To pack a column with 1 ml resin bed volume, 2 ml 50% suspension are necessary.

Specifications

Binding Capacity Biotin Binding Capacity > 40 nmol/ml resin
Capacity (Metric) 4 mL
Formulation 50% suspension in buffer, pH 8: 100 mM Tris-HCl pH 8.0, 1 mM EDTA, 150 mM NaCl, 0.01% sodium azide
Support Type Crosslinked agarose
For Use With (Application) For Packing of Gravity Flow Columns for Affinity Chromatographic Cell or Exosome Isolation

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.