Learn More
IBA Lifesciences Strep-Tactin™ PE or APC
GREENER_CHOICE
Product Code. 15653888 Shop All IBA Lifesciences Products
Click to view available options
Label or Dye:
APC Conjugate
PE Conjugate
Unit Size:
50µL
Product Code. 15653888
Brand: IBA Lifesciences 65000001
Fluorophore-conjugated Strep-Tactin™ for cell staining with PE or APC
  • Strep-Tactin™ conjugated to either R-Phycoerythrin (PE) or Allophycocyanin (APC) is suitable for staining or isolation of antigen-specific cytotoxic T cells (MHC I Streptamer™) or surface marker-specific cells (Fab-TACS™/Nano-TACS™) based on IBAs Strep-tag™ technology.
  • To target specific cells, MHC I complexes, low affinity Fab fragments or nanobodies fused to a Twin-Strep-tag™ (MHC I-Streps, Fab-Streps or Nano-Streps) are required in addition to fluorescent Strep-Tactin™.
  • Strep-Tactin™ multimerizes MHC I-Streps or Fab-/Nano-Streps, ensuring stable binding to cells.
  • The resulting fluorescent complexes can be used to stain cells that are specific for the respective peptide-loaded MHC I molecule or for a selected surface marker.
  • The addition of biotin reverses the multimerization and causes the dissociation of MHC I-Streps or Fab-/Nano-Streps from the cell surface.
  • The complete removal of all labeling reagents preserves the cells’ full biological activity.
Specifications
Kit Contents 50 tests a 1 to 2 x 106 cells
Quantity 50 Tests
Label or Dye PE Conjugate