missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Strep-Tactin™ 4Flow™ high capacity FPLC columns are designed for purifying Strep-tag™II and Twin-Strep-tag™ proteins via affinity chromatography in automated protein purification with HPLC/FPLC devices such as Äkta™ systems (Cytiva).
- Recommended for all protein classes due to the broad pH compatibility, excellent pressure stability, outstanding target purity (>95%), and high binding affinity
- 4% crosslinked agarose beads coupled to Strep-Tactin™ for straightforward purification of proteins with Strep-tag™II or a Twin-Strep-tag™
- Multiple regeneration cycles possible without loss of binding capacity
- Ideal alternative for proteins that are difficult to elute from Strep-Tactin™XT 4Flow™ resins
- Biotinylated proteins also bind to Strep-Tactin™ 4Flow™ high capacity and need to be blocked using BioLock
Specifications
Specifications
| Binding Capacity | 20 mg/mL resin |
| Capacity (Metric) | 1 mL |
| Description | 4% agarose, crosslinked |
| Particle Size | 50 to 150 μm |
| Form | Resin Slurry |
| Quantity | 5 mL |
| Protein Form | Tetrameric protein |
| Target | Proteins tagged with Twin-Strep-tag™; Strep-tag™II; cross reactivity: biotin |
| Type | Column |
| Final Product Type | Affinity chromatography equipment |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?