missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STRBP Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | STRBP |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18625830
|
Novus Biologicals
NBP2-94570-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18677281
|
Novus Biologicals
NBP2-94570-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
STRBP Polyclonal antibody specifically detects STRBP in Human samples. It is validated for Western BlotSpecifications
| STRBP | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DKFZp434N214, FLJ11307, FLJ14223, FLJ14984, ILF3L, interleukin enhancer binding factor 3-like, MGC21529, MGC3405, p74, spermatid perinuclear RNA binding protein, spermatid perinuclear RNA-binding protein, SPNRFLJ21960 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 40-190 of human STRBP (NP_060857.2). CALKHVSDWLDETNKGTKTEGETEVKKDEAGENYSKDQGGRTLCGVMRIGLVAKGLLIKDDMDLELVLMCKDKPTETLLNTVKDNLPIQIQKLTEEKYQVEQCVNEASIIIRNTKEPTLTLKVILTSPLIRDELEKKDGENVSMKDPPDLL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 55342 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title