missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STRBP Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94570-0.1ml
This item is not returnable.
View return policy
Description
STRBP Polyclonal antibody specifically detects STRBP in Human samples. It is validated for Western Blot
Specifications
| STRBP | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DKFZp434N214, FLJ11307, FLJ14223, FLJ14984, ILF3L, interleukin enhancer binding factor 3-like, MGC21529, MGC3405, p74, spermatid perinuclear RNA binding protein, spermatid perinuclear RNA-binding protein, SPNRFLJ21960 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 40-190 of human STRBP (NP_060857.2). CALKHVSDWLDETNKGTKTEGETEVKKDEAGENYSKDQGGRTLCGVMRIGLVAKGLLIKDDMDLELVLMCKDKPTETLLNTVKDNLPIQIQKLTEEKYQVEQCVNEASIIIRNTKEPTLTLKVILTSPLIRDELEKKDGENVSMKDPPDLL | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 55342 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction