missing translation for 'onlineSavingsMsg'
Learn More

Cerillo™ Stratus Plate Reader

Product Code. 17331819
Encompass_Preferred
Click to view available options
Wavelength Range:
496 nm
600 nm
650 nm
Custom
Packaging:
1 Ea.
4/Pk.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17331819

Brand: Cerillo™ 4PR2600

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The Stratus is small enough to fit in the palm of your hand and allows for measurements in small spaces or restrictive environments, like anaerobic or microaerophilic chambers and incubators. Its uniquely simple interface and data storage allow it to be placed anywhere, with or without a computer.

The Stratus plate reader has new and improved capabilities to optimize flexibility for sample data collection in multiple standard plate formats to serve the wide-ranging needs of scientific researchers. The maximum allowable shaking speed is determined by the shaking radius of the shaking platform used, as well as the method of adapter attachment.

  • Available in standard 600 nm, 496 nm, 650 nm, or customized wavelength
  • Fits standard height 6-, 12-, 24-, and 96-well plates
  • Small footprint and stackable
  • No moving parts; minimal maintenance
  • Functional in a broad range of environments
  • Data is stored on a microSD card, no need for direct connection to a computer
  • Can be powered by a wall outlet, external battery pack, or computer

Specifications

Detector Type Digital Photosensor
Dimensions (L x W x H) 14.4 x 13.0 x 6.8 cm
Incubator Temperature 50°C
Light Source Monochromatic Solid-State LEDs
Measurement Speed 45 sec.
Plate Type 6, 12, 24, and 96-well Plates
Product Line Stratus
Reading Options Kinetic
Software Type PC Software
Wavelength Selection Single Wavelength
Absorbance Range 0 to 3 OD
Accuracy <1% and 0.015 OD (0.000 to 2.000 OD)
Certifications/Compliance CE
CO2 Concentration Range 0 to 25%
Electrical Requirements 500 mA USB Supply
For Use With (Application) Absorbance based kinetic assays
Includes microSD card, power adapter, USB cable
Linearity <1% (0.000 to 2.000 OD)
O2 Concentration Range 0 to 21%
Packaging 4/Pk.
Precision <1% and 0.006 OD (0.000 to 2.000 OD)
Resolution 0.005 OD
Voltage 5 V
Warranty 2 Years
Wavelength Range 600 nm
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.