missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STRA13 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-55505
This item is not returnable.
View return policy
Description
STRA13 Polyclonal specifically detects STRA13 in Human samples. It is validated for Western Blot.
Specifications
| STRA13 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| BHLHE40, CENPX, CENP-X, DEC1, FAAP10, FANCM-interacting histone fold protein 2, Fanconi anemia-associated polypeptide of 10 kDa, HLHB2, MGC14480, MHF2centromere protein X, Retinoic acid-inducible gene D9 protein homolog, stimulated by retinoic acid 13, stimulated by retinoic acid 13 homolog (mouse), Stimulated by retinoic acid gene 13 protein homolog | |
| Rabbit | |
| 7 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| A8MT69-2 | |
| CENPX | |
| Synthetic peptides corresponding to STRA13(stimulated by retinoic acid 13 homolog (mouse)) The peptide sequence was selected from the middle region of STRA13. Peptide sequence MEGAGAGSGFRKELVSRLLHLHFKDDKTKEAAVRGVRQAQAEDALRVDVD. | |
| Affinity purified | |
| RUO | |
| 201254 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction