missing translation for 'onlineSavingsMsg'
Learn More

Wesbart™ Storage Bin

Product Code. 15755855
Click to view available options
Dimensions (L x W x H):
100 x 510 mm (HxW)
100 x 520 mm (HxW)
200 x 155 mm (HxW)
220 x 180 mm (HxW)
225 x 232 mm (HxW)
235 x 155 mm (HxW)
235 x 180 mm (HxW)
240 x 190 mm (HxW)
240 x 280 mm (HxW)
280 x 140 mm (HxW)
280 x 280 mm (HxW)
560 x 170 x 200 mm
600 x 180 x 280 mm
680 x 140 x 280 mm
90 x 133 mm (HxW)
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15755855

Brand: Wesbart™ BIN871

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

A useful way of keeping loose items safe within a freezer

  • Storage bins are manufactured from anodised aluminum or stainless steel
  • All storage bins have either a fold down handle or hand grip
  • Bins can be designed to have a handle on each end for transport if required
  • All storage bins are fitted with a label holder so the user can easily identify which bin they require
  • Specifications

    Type Storage Bin
    Material Anodized Aluminum, Stainless Steel
    For Use With Freezer
    For Use With (Application) Keeping Loose Items Safe within a Freezer
    Includes Handgrip and Label Holder
    Dimensions (L x W x H) 90 x 133 mm (HxW)
    Width (Metric) 133 mm
    Height (Metric) 90 mm
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.