Learn More
Eppendorf Storage Base for CellXpert CS220
Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
CellXpert CS220
Unit Size:
Each
Description
- Heaight: 65 cm
- For single device only
- Adjustable feet for levelling
- Allows easy cleaning below device and access with standard forklift to move devices
- Eppendorf service required for installation
Specifications
Specifications
| Product Type | Positioning Base |
| For Use With (Equipment) | CellXpert CS220 |