Learn More
Fisherbrand™ Fisherbrand BS 2775 Two Holed Rubber Stopper
Product Code. 10717321 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Red
Unit Size:
Pack of 5
Product Code. Color Unit Size
10717321 Red Pack of 5
1 options
Product Code. 10717321
  • ONLINE EXCLUSIVE PRICE
    £28.63
    £24.00 / Pack of 5
    Save £4.63
    16% Off
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
Prevents liquid chemical leaks or escape from container.
  • Small, tapered plug used to seal the openings of test tubes, flasks and other laboratory glassware
  • Preferable for applications that require tighter seal or greater degree of chemical resistance
  • Prevent contamination of samples by protecting contents of laboratory glassware from air
  • Allows experimenters to shake or mix solutions without spilling them
Specifications
Description Two holed
Quantity 5/Pk.
Color Red
Material Rubber
Product Type Tapered Stopper (2 holes)