Learn More
Invitrogen™ Human STK38L (NDR2), GST Tag Recombinant Protein
Human STK38L (NDR2), GST Tag Recombinant Protein
Brand: Invitrogen™ PV6374
Code : NEW
Additional Details : Weight : 0.01000kg
Description
STK38L act as novel pro-apoptotic kinase that is activated by RASSF1A/MST1 in response to Fas receptor stimulation and promotes apoptosis. STK38L is mainly involved in the regulation of structural processes in differentiating and mature neuronal cells and it is highly expressed with highest levels observed in the thymus.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation | |
80.3kDa | |
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. | |
Human | |
GST Tag |
50mM Tris-HCl pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol | |
100μg | |
KIAA0965, STK38L | |
Recombinant |