missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human STK38L (NDR2), GST Tag Recombinant Protein
Human STK38L (NDR2), GST Tag Recombinant Protein
£431.00 - £6530.00
Specifications
Gene Alias | KIAA0965, STK38L |
---|---|
Species | Human |
Recombinant | Recombinant |
Protein Tag | GST Tag |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15441224
|
Invitrogen™
PV6373 |
10μg |
£431.00
10µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15489003
|
Invitrogen™
PV6375 |
1mg |
£6530.00
1mg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15479003
|
Invitrogen™
PV6374 |
100μg |
£1460.00
100µg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
STK38L act as novel pro-apoptotic kinase that is activated by RASSF1A/MST1 in response to Fas receptor stimulation and promotes apoptosis. STK38L is mainly involved in the regulation of structural processes in differentiating and mature neuronal cells and it is highly expressed with highest levels observed in the thymus.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
KIAA0965, STK38L | |
Human | |
Recombinant | |
GST Tag |