missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK24 Mouse, anti-Human, Clone: 2B4C1, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | STK24 |
|---|---|
| Clone | 2B4C1 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18000316
|
Proteintech
68522-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18094765
|
Proteintech
68522-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The STK24 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human STK24. This antibody recognizes human, mouse, rat antigen. The STK24 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| STK24 | |
| 1000 μg/mL | |
| Liquid | |
| Human, Mouse, Rat | |
| MST 3, MST3, MST3B, STE20, STE20 like kinase MST3, STK24, STK3 | |
| Primary |
| 2B4C1 | |
| Western Blot, ELISA | |
| Mouse | |
| 8428 | |
| IgG1 | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title