Learn More
STK24 Mouse, anti-Human, Clone: 2B4C1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68522-1-IG-20UL
This item is not returnable.
View return policy
Description
The STK24 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human STK24. This antibody recognizes human, mouse, rat antigen. The STK24 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| STK24 | |
| 2B4C1 | |
| MST 3, MST3, MST3B, STE20, STE20 like kinase MST3, STK24, STK3 | |
| 20 μL | |
| 8428 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG1 |