missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Magnetic Stirring Bar with Pivot Ring

Create high turbulence even at a low speed with a stirring bar that features and angled surface. Kartell™ Magnetic Stirring Bar with Pivot Ring reduces the contact surface and fosters optimal stir bar positioning.

Brand:  Kartell™ 0078800

90.68 GBP valid until 2024-04-30
Use promo code "21656" to get your promotional price.



Code : A0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10274281

  • £95.45 / Pack of 10
Estimated Shipment: 29-05-2024
to see stock.
Add to basket

Description

Description

Create high turbulence even at a low speed with a stirring bar that features and angled surface. Kartell™ Magnetic Stirring Bar with Pivot Ring reduces the contact surface and fosters optimal stir bar positioning.

  • Angled surfaces create high turbulence even at low speed
  • For containers with irregular bottom
  • The pivot ring reduces the contact surface
  • Suitable for foodstuff
Specifications

Specifications

Pivot ring
Magnet PTFE Coated
10/Pk.
35.48 mm
6.53 mm
White
PTFE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers